Assay Details
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019375.1 | NP_062248.1 | 2 - 3 | 235 | 111 | |
GenBank mRNA | AF111179.1 | - | 2 - 3 | 235 | 111 | |
AF111181.1 | - | 2 - 3 | 234 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
