Assay Details
Gene Symbol: Cpsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106785.1 | NP_001100255.1 | 1 - 2 | 102 | 137 | |
XM_006241372.1 | XP_006241434.1 | 1 - 2 | 134 | 137 | ||
XM_006241373.1 | XP_006241435.1 | 1 - 2 | 134 | 137 | ||
XM_006241374.1 | XP_006241436.1 | 1 - 2 | 134 | 137 | ||
XM_008765386.1 | XP_008763608.1 | 1 - 2 | 134 | 137 | ||
XM_008765387.1 | XP_008763609.1 | 1 - 2 | 134 | 137 | ||
XM_008765388.1 | XP_008763610.1 | 1 - 2 | 134 | 137 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106785.1, XM_006241372.1, XM_006241373.1, XM_006241374.1, XM_008765386.1, XM_008765387.1, XM_008765388.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
