Assay Details
Gene Symbol: Cpsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106785.1 | NP_001100255.1 | 2 - 3 | 306 | 145 | |
XM_006241372.1 | XP_006241434.1 | 2 - 3 | 338 | 145 | ||
XM_006241373.1 | XP_006241435.1 | 2 - 3 | 338 | 145 | ||
XM_006241374.1 | XP_006241436.1 | 2 - 3 | 338 | 145 | ||
XM_008765386.1 | XP_008763608.1 | 2 - 3 | 338 | 145 | ||
XM_008765387.1 | XP_008763609.1 | 2 - 3 | 338 | 145 | ||
XM_008765388.1 | XP_008763610.1 | 2 - 3 | 338 | 145 | ||
XM_008765390.1 | XP_008763612.1 | 2 - 3 | 263 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
