Assay Details
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021843.4 | NP_068615.1 | 6 - 7 | 783 | 87 | |
XM_008765339.1 | XP_008763561.1 | 6 - 7 | 827 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
