Assay Details
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053561.2 | NP_446013.2 | 12 - 13 | 1214 | 96 | |
XM_006241342.1 | XP_006241404.1 | 12 - 13 | 1228 | 96 | ||
XM_006241343.2 | XP_006241405.1 | 12 - 13 | 1228 | 96 | ||
XM_006241344.2 | XP_006241406.1 | 10 - 11 | 965 | 96 | ||
GenBank mRNA | AF062594.1 | - | 12 - 13 | 1196 | 96 | |
CF110129.1 | - | 7 - 8 | 649 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
