Assay Details
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053561.2 | NP_446013.2 | 1 - 2 | 142 | 107 | |
XM_006241342.1 | XP_006241404.1 | 1 - 2 | 156 | 107 | ||
XM_006241343.2 | XP_006241405.1 | 1 - 2 | 156 | 107 | ||
GenBank mRNA | CO574484.1 | - | 158 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_053561.2, XM_006241342.1, XM_006241343.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
