Assay Details
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053561.2 | NP_446013.2 | 6 - 7 | 597 | 117 | |
XM_006241342.1 | XP_006241404.1 | 6 - 7 | 611 | 117 | ||
XM_006241343.2 | XP_006241405.1 | 6 - 7 | 611 | 117 | ||
XM_006241344.2 | XP_006241406.1 | 4 - 5 | 348 | 117 | ||
GenBank mRNA | AF062594.1 | - | 6 - 7 | 582 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
