Assay Details
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001056437.5 | XP_001056437.2 | 5 - 6 | 2167 | 99 | |
XM_006241237.2 | XP_006241299.1 | 5 - 6 | 495 | 99 | ||
XM_008765305.1 | XP_008763527.1 | 5 - 6 | 2951 | 99 | ||
XM_008765306.1 | XP_008763528.1 | 6 - 7 | 2482 | 99 | ||
XM_008765307.1 | XP_008763529.1 | 6 - 7 | 3037 | 99 | ||
XM_008765308.1 | XP_008763530.1 | 6 - 7 | 2485 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
