Assay Details
Gene Symbol: LOC100910056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750307.3 | XP_003750355.3 | 4 - 5 | 1975 | 132 |
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108753.1 | NP_001102223.1 | 17 - 18 | 3951 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
