Assay Details
Gene Symbol: Arl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022385.3 | NP_071780.1 | 2 - 3 | 238 | 104 | |
GenBank mRNA | BC061553.1 | - | 321 | 104 | ||
FQ226660.1 | - | 2 - 3 | 294 | 104 | ||
U12402.1 | - | 2 - 3 | 170 | 104 | ||
X76920.1 | - | 2 - 3 | 238 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
