Assay Details
Gene Symbol: Glt8d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765296.1 | XP_008763518.1 | 4 - 5 | 958 | 112 | |
XM_008765297.1 | XP_008763519.1 | 4 - 5 | 957 | 112 | ||
XM_008765298.1 | XP_008763520.1 | 4 - 5 | 957 | 112 | ||
XM_008765299.1 | XP_008763521.1 | 4 - 5 | 957 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
