Assay Details
Gene Symbol: Frs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108097.3 | NP_001101567.2 | 5 - 6 | 417 | 74 | |
XM_006241381.2 | XP_006241443.1 | 3 - 4 | 359 | 74 | ||
GenBank mRNA | CB581691.1 | - | 3 - 4 | 240 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108097.3, XM_006241381.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
