Assay Details
Gene Symbol: Frs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108097.3 | NP_001101567.2 | 9 - 10 | 1017 | 78 | |
XM_006241381.2 | XP_006241443.1 | 7 - 8 | 959 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
gastrulation with mouth forming second
organ induction
lens development in camera-type eye
ventricular septum development
transmembrane receptor protein tyrosine kinase signaling pathway
neuroblast proliferation
fibroblast growth factor receptor signaling pathway
anterior/posterior axis specification, embryo
positive regulation of signal transduction
forebrain development
regulation of apoptotic process
lens placode formation involved in camera-type eye formation
regulation of epithelial cell proliferation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
lens fiber cell development
regulation of ERK1 and ERK2 cascade
transmembrane receptor protein tyrosine kinase adaptor activity
fibroblast growth factor receptor binding
insulin receptor binding
neurotrophin TRKA receptor binding
gastrulation with mouth forming second
organ induction
lens development in camera-type eye
ventricular septum development
transmembrane receptor protein tyrosine kinase signaling pathway
neuroblast proliferation
fibroblast growth factor receptor signaling pathway
anterior/posterior axis specification, embryo
positive regulation of signal transduction
forebrain development
regulation of apoptotic process
lens placode formation involved in camera-type eye formation
regulation of epithelial cell proliferation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
lens fiber cell development
regulation of ERK1 and ERK2 cascade
transmembrane receptor protein tyrosine kinase adaptor activity
fibroblast growth factor receptor binding
insulin receptor binding
neurotrophin TRKA receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P663544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P663544 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P663544 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P663544 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P449422 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P548740 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20936 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29465 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53423 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10902528 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902526 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902529 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902527 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902530 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902525 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10902524 | - | - |
Rat Genome 230 2.0 Array | 1397648_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397648_at | - | - |
Rat Genome 230 2.0 Array | 1377661_at | - | - |
Rat Genome 230 2.0 Array | 1377661_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1397648_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377661_at | - | - |
Affymetrix HT Rat Focus Array | 1397648_at | - | - |
Affymetrix HT Rat Focus Array | 1377661_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700021393.rn.1 | TC0700002748.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021392.rn.1 | TC0700002748.rn.1 | RTA TC |
Rat Expression Set 230 | 1397648_at | - | - |
Rat Expression Set 230 | 1377661_PM_at | - | - |
Rat Expression Set 230 | 1377661_at | - | - |
Rat Expression Set 230 | 1397648_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902524 | - | - |
Rat Genome U34 Set | rc_AA899046_at | - | - |
Rat Genome U34 Set | rc_AA996518_at | - | - |
Rat Genome U34 Set | rc_AI233853_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21075243 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373764 | - | - |
