Assay Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241191.2 | XP_006241253.1 | 1 - 2 | 94 | 77 | |
XM_006241193.2 | XP_006241255.1 | 1 - 2 | 93 | 77 | ||
GenBank mRNA | FQ209824.1 | - | 1 - 2 | 82 | 77 | |
X06108.1 | - | 1 - 2 | 42 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X06108.1, XM_006241191.2, XM_006241193.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
