Assay Details
Gene Symbol: Tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053729.1 | NP_446181.1 | 6 - 7 | 736 | 94 | |
GenBank mRNA | AY026945.1 | - | 6 - 7 | 736 | 94 | |
BC129088.1 | - | 6 - 7 | 753 | 8362408 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
