Assay Details
Gene Symbol: Rab21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004238.1 | NP_001004238.1 | 5 - 6 | 658 | 127 | |
GenBank mRNA | BC079323.1 | - | 5 - 6 | 658 | 127 | |
FQ226410.1 | - | 4 - 5 | 260 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
