Assay Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241616.1 | XP_006241678.1 | 1 - 2 | 106 | 153 | |
XM_006241617.1 | XP_006241679.1 | 1 - 2 | 106 | 153 | ||
GenBank mRNA | FQ211706.1 | - | 1 - 2 | 106 | 153 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241616.1, XM_006241617.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
