Assay Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241616.1 | XP_006241678.1 | 4 - 5 | 595 | 94 | |
XM_006241617.1 | XP_006241679.1 | 4 - 5 | 595 | 94 | ||
XM_006241618.2 | XP_006241680.1 | 3 - 4 | 755 | 94 | ||
XM_008765476.1 | XP_008763698.1 | 3 - 4 | 536 | 94 | ||
XM_008765477.1 | XP_008763699.1 | 3 - 4 | 539 | 94 | ||
GenBank mRNA | FQ211706.1 | - | 4 - 5 | 595 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
