Assay Details
Gene Symbol: Tac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019162.2 | NP_062035.1 | 6 - 7 | 485 | 154 | |
GenBank mRNA | FQ224503.1 | - | 3 - 4 | 227 | 154 | |
M16410.1 | - | 6 - 7 | 485 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
