Assay Details
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U62316.1 | - | 1 - 2 | 169 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U62316.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
