Assay Details
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017302.2 | NP_058998.2 | 3 - 4 | 442 | 75 | |
XM_006241432.2 | XP_006241494.1 | 4 - 5 | 984 | 75 | ||
XM_006241433.2 | XP_006241495.1 | 5 - 6 | 2500 | 75 | ||
XM_006241434.2 | XP_006241496.1 | 5 - 6 | 851 | 75 | ||
XM_006241435.2 | XP_006241497.1 | 4 - 5 | 2464 | 75 | ||
XM_006241436.2 | XP_006241498.1 | 4 - 5 | 726 | 75 | ||
XM_006241437.2 | XP_006241499.1 | 4 - 5 | 2374 | 75 | ||
GenBank mRNA | BC081701.1 | - | 3 - 4 | 442 | 75 | |
FQ218176.1 | - | 4 - 5 | 556 | 75 | ||
U62316.1 | - | 4 - 5 | 608 | 75 | ||
X97445.1 | - | 4 - 5 | 567 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
