Assay Details
Gene Symbol: Il2rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013195.1 | NP_037327.1 | 3 - 4 | 89 | ||
XM_006241970.2 | XP_006242032.1 | 3 - 4 | 747 | 89 | ||
GenBank mRNA | M55050.1 | - | 3 - 4 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
