Assay Details
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024398.2 | NP_077374.2 | 2 - 3 | 181 | 103 | |
GenBank mRNA | AJ243266.1 | - | 2 - 3 | 182 | 103 | |
BC061999.1 | - | 2 - 3 | 181 | 103 | ||
FQ216755.1 | - | 2 - 3 | 206 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
