Assay Details
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241900.2 | XP_006241962.1 | 7 - 8 | 915 | 67 | |
XM_006241901.2 | XP_006241963.1 | 7 - 8 | 916 | 67 | ||
XM_006241902.1 | XP_006241964.1 | 5 - 6 | 529 | 67 | ||
XM_008765628.1 | XP_008763850.1 | 7 - 8 | 2304 | 67 | ||
XM_216954.8 | XP_216954.4 | 7 - 8 | 914 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
