Assay Details
Gene Symbol: Eif3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011990.1 | NP_001011990.1 | 2 - 3 | 211 | 144 | |
XM_006250385.2 | XP_006250447.1 | 2 - 3 | 253 | 144 | ||
GenBank mRNA | BC082087.1 | - | 2 - 3 | 211 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
