Assay Details
Gene Symbol: Eif3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011990.1 | NP_001011990.1 | 3 - 4 | 329 | 65 | |
XM_006250385.2 | XP_006250447.1 | 3 - 4 | 371 | 65 | ||
GenBank mRNA | BC082087.1 | - | 3 - 4 | 329 | 65 | |
FQ215133.1 | - | 1 - 2 | 140 | 65 | ||
FQ216598.1 | - | 2 - 3 | 140 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
