Assay Details
Gene Symbol: Eif3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011990.1 | NP_001011990.1 | 6 - 7 | 605 | 87 | |
XM_006250385.2 | XP_006250447.1 | 6 - 7 | 647 | 87 | ||
GenBank mRNA | BC082087.1 | - | 6 - 7 | 605 | 7133833 | 87 |
FQ216598.1 | - | 5 - 6 | 416 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
