Assay Details
Gene Symbol: Taf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133319.1 | NP_579853.1 | 11 - 12 | 1627 | 166 | |
XM_006241644.1 | XP_006241706.1 | 7 - 8 | 941 | 166 | ||
GenBank mRNA | BC105765.1 | - | 11 - 12 | 1627 | 166 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
