Assay Details
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130559.1 | NP_001124031.1 | 5 - 6 | 1050 | 150 | |
XM_006241544.2 | XP_006241606.1 | 5 - 6 | 882 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
