Assay Details
Gene Symbol: Slc25a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126269.1 | NP_001119741.1 | 1 - 2 | 178 | 81 | |
GenBank mRNA | BC166457.1 | - | 1 - 2 | 73 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
