Assay Details
Gene Symbol: Rrm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130543.1 | NP_001124015.1 | 4 - 5 | 588 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
