Assay Details
Gene Symbol: Gpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031039.1 | NP_112301.1 | 1 - 2 | 276 | 68 | |
XM_008765599.1 | XP_008763821.1 | 2 - 3 | 996 | 68 | ||
GenBank mRNA | BC085728.1 | - | 1 - 2 | 260 | 68 | |
BC097937.1 | - | 1 - 2 | 280 | 68 | ||
D10354.1 | - | 1 - 2 | 276 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
