Assay Details
Gene Symbol: Gpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031039.1 | NP_112301.1 | 6 - 7 | 929 | 81 | |
XM_008765599.1 | XP_008763821.1 | 7 - 8 | 1649 | 81 | ||
GenBank mRNA | BC097937.1 | - | 6 - 7 | 933 | 81 | |
D10354.1 | - | 6 - 7 | 929 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
