Assay Details
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109976.2 | NP_001103446.1 | 7 - 8 | 787 | 93 | |
GenBank mRNA | DY311072.1 | - | 2 - 3 | 131 | 93 | |
EV771639.1 | - | 5 - 6 | 559 | 94 | ||
FQ211519.1 | - | 7 - 8 | 1222 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
