Assay Details
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130504.1 | NP_001123976.1 | 5 - 6 | 470 | 79 | |
XM_008765720.1 | XP_008763942.1 | 4 - 5 | 632 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
