Assay Details
Gene Symbol: Rpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198753.2 | NP_942048.1 | 5 - 6 | 712 | 140 | |
GenBank mRNA | BC058494.1 | - | 5 - 6 | 715 | 6887923 | 140 |
FQ209477.1 | - | 5 - 6 | 711 | 140 | ||
FQ211542.1 | - | 5 - 6 | 712 | 140 | ||
FQ212671.1 | - | 5 - 6 | 140 | |||
FQ213135.1 | - | 5 - 6 | 714 | 140 | ||
FQ214177.1 | - | 5 - 6 | 712 | 140 | ||
FQ214687.1 | - | 5 - 6 | 709 | 140 | ||
FQ218147.1 | - | 5 - 6 | 712 | 140 | ||
FQ219083.1 | - | 5 - 6 | 712 | 140 | ||
FQ219302.1 | - | 5 - 6 | 712 | 140 | ||
FQ219523.1 | - | 5 - 6 | 711 | 140 | ||
FQ220029.1 | - | 5 - 6 | 711 | 140 | ||
FQ220099.1 | - | 5 - 6 | 712 | 140 | ||
FQ220378.1 | - | 5 - 6 | 712 | 140 | ||
FQ220408.1 | - | 5 - 6 | 708 | 140 | ||
FQ220423.1 | - | 5 - 6 | 711 | 140 | ||
FQ222720.1 | - | 5 - 6 | 712 | 140 | ||
FQ222993.1 | - | 5 - 6 | 712 | 140 | ||
FQ223514.1 | - | 5 - 6 | 712 | 140 | ||
FQ225290.1 | - | 5 - 6 | 712 | 140 | ||
FQ225371.1 | - | 2 - 3 | 296 | 140 | ||
FQ225405.1 | - | 5 - 6 | 712 | 140 | ||
FQ225635.1 | - | 5 - 6 | 712 | 140 | ||
FQ227897.1 | - | 5 - 6 | 712 | 140 | ||
FQ228916.1 | - | 5 - 6 | 712 | 140 | ||
FQ229153.1 | - | 5 - 6 | 711 | 139 | ||
FQ229505.1 | - | 5 - 6 | 712 | 140 | ||
FQ229795.1 | - | 5 - 6 | 708 | 140 | ||
FQ229869.1 | - | 5 - 6 | 712 | 140 | ||
FQ229906.1 | - | 5 - 6 | 712 | 140 | ||
FQ230150.1 | - | 5 - 6 | 712 | 140 | ||
FQ231551.1 | - | 5 - 6 | 708 | 140 | ||
FQ232038.1 | - | 5 - 6 | 712 | 140 | ||
FQ232351.1 | - | 3 - 4 | 454 | 140 | ||
FQ233196.1 | - | 5 - 6 | 712 | 140 | ||
FQ234897.1 | - | 5 - 6 | 712 | 140 | ||
X62166.1 | - | 5 - 6 | 716 | 140 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
