Assay Details
Gene Symbol: Card10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130554.1 | NP_001124026.1 | 14 - 15 | 2090 | 67 | |
XM_006241986.2 | XP_006242048.1 | 14 - 15 | 2216 | 67 | ||
XM_006241987.2 | XP_006242049.1 | 14 - 15 | 2216 | 67 | ||
XM_006241988.2 | XP_006242050.1 | 14 - 15 | 3131 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
