Assay Details
Gene Symbol: Grap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034944.1 | NP_001030116.1 | 3 - 4 | 295 | 74 | |
GenBank mRNA | BC105895.1 | - | 3 - 4 | 295 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
