Assay Details
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241729.2 | XP_006241791.1 | 2 - 3 | 195 | 68 | |
XM_008765573.1 | XP_008763795.1 | 2 - 3 | 1383 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241729.2, XM_008765573.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
