Assay Details
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114405.2 | NP_001107877.1 | 3 - 4 | 823 | 86 | |
XM_006241729.2 | XP_006241791.1 | 5 - 6 | 1320 | 86 | ||
XM_006241731.2 | XP_006241793.1 | 4 - 5 | 2430 | 86 | ||
XM_006241732.2 | XP_006241794.1 | 4 - 5 | 1242 | 86 | ||
XM_006241733.2 | XP_006241795.1 | 4 - 5 | 1201 | 86 | ||
XM_008765573.1 | XP_008763795.1 | 5 - 6 | 2508 | 86 | ||
XM_008765574.1 | XP_008763796.1 | 4 - 5 | 1887 | 86 | ||
GenBank mRNA | BC159432.1 | - | 3 - 4 | 823 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
