Assay Details
Gene Symbol: Sqle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017136.2 | NP_058832.1 | 1 - 2 | 354 | 109 | |
GenBank mRNA | BC097330.1 | - | 1 - 2 | 354 | 7460860 | 109 |
D37920.1 | - | 1 - 2 | 485 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
