Assay Details
Gene Symbol: Cyp27b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053763.1 | NP_446215.1 | 5 - 6 | 969 | 98 | |
XM_008765374.1 | XP_008763596.1 | 6 - 7 | 2841 | 98 | ||
GenBank mRNA | AB001992.1 | - | 5 - 6 | 969 | 98 | |
AF000139.1 | - | 5 - 6 | 961 | 98 |
Gene Symbol: LOC100909462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773774.1 | XP_008771996.1 | 1729 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
