Assay Details
Gene Symbol: Cdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053593.2 | NP_446045.1 | 3 - 4 | 544 | 68 | |
GenBank mRNA | BC070956.1 | - | 3 - 4 | 544 | 68 | |
FQ214013.1 | - | 3 - 4 | 560 | 68 | ||
FQ219275.1 | - | 3 - 4 | 520 | 68 | ||
FQ234663.1 | - | 2 - 3 | 173 | 68 | ||
L11007.1 | - | 3 - 4 | 476 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
