Assay Details
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012200.1 | NP_001012200.1 | 5 - 6 | 582 | 108 | |
XM_006242102.2 | XP_006242164.1 | 5 - 6 | 587 | 108 | ||
XM_006242103.2 | XP_006242165.1 | 5 - 6 | 633 | 108 | ||
XM_006242104.1 | XP_006242166.1 | 4 - 5 | 628 | 108 | ||
XM_008765764.1 | XP_008763986.1 | 4 - 5 | 539 | 108 | ||
GenBank mRNA | BC087669.1 | - | 5 - 6 | 582 | 7316813 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
