Assay Details
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012200.1 | NP_001012200.1 | 6 - 7 | 756 | 94 | |
XM_006242102.2 | XP_006242164.1 | 6 - 7 | 761 | 94 | ||
XM_006242103.2 | XP_006242165.1 | 6 - 7 | 807 | 94 | ||
XM_006242104.1 | XP_006242166.1 | 5 - 6 | 802 | 94 | ||
XM_008765764.1 | XP_008763986.1 | 5 - 6 | 713 | 94 | ||
GenBank mRNA | BC087669.1 | - | 6 - 7 | 756 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
