Assay Details
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130566.1 | NP_001124038.1 | 3 - 4 | 1142 | 77 | |
XM_008765580.1 | XP_008763802.1 | 2 - 3 | 3616 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
