Assay Details
Gene Symbol: Mrpl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241651.1 | XP_006241713.1 | 1 - 2 | 228 | 80 | |
GenBank mRNA | FQ220388.1 | - | 1 - 2 | 214 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241651.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
