Assay Details
Gene Symbol: Rspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130575.1 | NP_001124047.1 | 2 - 3 | 1283 | 150 | |
XM_008765454.1 | XP_008763676.1 | 2 - 3 | 1433 | 150 | ||
XM_008765455.1 | XP_008763677.1 | 2 - 3 | 732 | 150 | ||
GenBank mRNA | AB232173.1 | - | 2 - 3 | 1435 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
