Assay Details
Gene Symbol: Ext1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130540.1 | NP_001124012.1 | 10 - 11 | 2175 | 76 | |
GenBank mRNA | BC105839.1 | - | 10 - 11 | 1490 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
ossification
glycosaminoglycan biosynthetic process
gastrulation
axon guidance
brain development
endoderm development
mesoderm development
biological_process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
olfactory bulb development
cellular polysaccharide biosynthetic process
embryonic skeletal joint development
molecular_function
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
glycosyltransferase activity
protein homodimerization activity
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity
ossification
glycosaminoglycan biosynthetic process
gastrulation
axon guidance
brain development
endoderm development
mesoderm development
biological_process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
olfactory bulb development
cellular polysaccharide biosynthetic process
embryonic skeletal joint development
molecular_function
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
glycosyltransferase activity
protein homodimerization activity
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158848 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158848 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158848 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158848 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P164068 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P183137 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10470 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86569 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87570 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10035 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4792 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12612 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85991 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81489 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85734 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42418 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56561 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2625 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15194 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36957 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10903710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903713 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903715 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903714 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903720 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903709 | - | - |
Rat Genome 230 2.0 Array | 1392083_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373393_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373393_at | - | - |
Rat Genome 230 2.0 Array | 1392083_at | - | - |
Affymetrix HT Rat Focus Array | 1392083_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373393_at | - | - |
Affymetrix HT Rat Focus Array | 1373393_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392083_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700023958.rn.1 | TC0700003127.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023957.rn.1 | TC0700003127.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023959.rn.1 | TC0700003127.rn.1 | RTA TC |
Rat Expression Set 230 | 1373393_at | - | - |
Rat Expression Set 230 | 1373393_PM_at | - | - |
Rat Expression Set 230 | 1392083_at | - | - |
Rat Expression Set 230 | 1392083_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903709 | - | - |
Rat Genome U34 Set | rc_AA858704_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21515975 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364547 | - | - |
RatRef-12 Expression BeadChip | ILMN_68463 | - | - |
