Assay Details
Gene Symbol: Ext1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130540.1 | NP_001124012.1 | 4 - 5 | 1406 | 152 | |
GenBank mRNA | BC105839.1 | - | 4 - 5 | 721 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
